Order Entry
Export
Orders LinkContactUsLinkComponent
 

1,3-Benzenedithiol

Bottle, 5 g
Catalog # APOSOR55264-5G
Corrosive
Irritant
Supplier:   Apollo Scientific
1,3-Benzenedithiol
Catalog # APOSOR55264-5G
Supplier:  Apollo Scientific
CAS Number:  626-04-0
1,3-Benzenedithiol

Bottle, 5 g

CAS Number:  626-04-0
Price
362,00 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  Benzene-1,3-dithiol
  • CAS Number:626-04-0
  • Formula:C₆H₆S₂
  • MDL Number:MFCD00004842
  • Molecular Weight:142.25 g/mol
  • Boiling Pt:244 °C
  • Melting Pt:24…25 °C
  • Density:1.24 g/cm³ (25 °C)
  • Flash Pt:113 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g

Specifications