Order Entry
Export
Orders LinkContactUsLinkComponent
1,4-Dimethylpyrazole

1,4-Dimethylpyrazole

Bottle, 25 g
Catalog # APOSOR350149-25G
Supplier:   Apollo Scientific
1,4-Dimethylpyrazole
Catalog # APOSOR350149-25G
Supplier:  Apollo Scientific
CAS Number:  1072-68-0
1,4-Dimethylpyrazole

Bottle, 25 g

CAS Number:  1072-68-0
Price
30,40 €
Log in for availability
1 * 25 g
Product Details & Documents
  • CAS Number:1072-68-0
  • Formula:C₅H₈N₂
  • MDL Number:MFCD01075158
  • Molecular Weight:96.1300 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g

Specifications