Order Entry
Export
Orders LinkContactUsLinkComponent
2,2-Difluoroacetamide 97%

2,2-Difluoroacetamide 97%

Bottle, 25 g
Catalog # APOSPC4051-25G
Irritant
Supplier:   Apollo Scientific
2,2-Difluoroacetamide 97%
Catalog # APOSPC4051-25G
Supplier:  Apollo Scientific
CAS Number:  359-38-6
2,2-Difluoroacetamide 97%

Bottle, 25 g

CAS Number:  359-38-6
Price
145,00 €
Log in for availability
1 * 25 g
Product Details & Documents
  • CAS Number:359-38-6
  • Formula:C₂H₃F₂NO
  • MDL Number:MFCD01896431
  • Molecular Weight:95.05 g/mol
  • Melting Pt:45…55 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g

Specifications