Order Entry
Export
Orders LinkContactUsLinkComponent
 

1,3-Cyclopentanedione

Bottle, 25 g
Catalog # APOSOR24952-25G
Irritant
Supplier:   Apollo Scientific
1,3-Cyclopentanedione
Catalog # APOSOR24952-25G
Supplier:  Apollo Scientific
CAS Number:  3859-41-4
1,3-Cyclopentanedione

Bottle, 25 g

CAS Number:  3859-41-4
Price
75,30 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  Cyclopentane-1,3-dione
  • CAS Number:3859-41-4
  • Formula:C₅H₆O₂
  • MDL Number:MFCD00001405
  • Molecular Weight:98.1 g/mol
  • Melting Pt:148…153 °C
  • Storage Temperature:Refrigerator
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g

Specifications