Orders LinkContactUsLinkComponent
SeramunBlock
SeramunBlock
SeramunBlock, 250 mL
Catalog # APOSBIL3122-250ML
Supplier:   Apollo Scientific
SeramunBlock
Catalog # APOSBIL3122-250ML
Supplier:  Apollo Scientific
CAS Number:  
SeramunBlock
SeramunBlock, 250 mL
CAS Number:  
Price
98,40 €
Log in for availability
1 * 250 mL
Product Details & Documents
Combined Stabiliser/ blocking solution for solid phase immobilised proteins effecting protein stabilisation and blocking of free surfaces.
Specifications
Specifications
  • Pk:
    250 mL
  • Description:
    SeramunBlock
Specifications