2,4-Dihydroxybenzaldehyde
Bottle, 500 g
Catalog # APOSOR1169-500G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
- CAS Number:95-01-2
- Formula:C₇H₆O₃
- MDL Number:MFCD00011686
- Molecular Weight:138.12 g/mol
- Boiling Pt:220 °C
- Melting Pt:134…138 °C
- Storage Temperature:Ambient
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Bottle
- Pk:500 g



