0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
1,3-Benzenedithiol
1,3-Benzenedithiol
1,3-Benzenedithiol
1,3-Benzenedithiol
Bottle, 1 g
APOSOR55264-1G
:
Apollo Scientific
1,3-Benzenedithiol
APOSOR55264-1G
:
Apollo Scientific
:
626-04-0
Expand image
1,3-Benzenedithiol
Bottle, 1 g
Add to cart
:
626-04-0
94,10 €
Unit of Measure
1 * 1 g
-
+
Add to cart
:
Benzene-1,3-dithiol
:
626-04-0
:
C₆H₆S₂
:
MFCD00004842
:
142.25 g/mol
:
244 °C
:
24…25 °C
:
1.24 g/cm³ (25 °C)
:
113 °C
:
Ambient
Pack type:
Bottle
Pk:
1 g