Order Entry
Export
Orders LinkContactUsLinkComponent
 

Cinnamaldehyde 98%

Bottle, 100 g
Catalog # APOSOR55404-100G
Irritant
Supplier:   Apollo Scientific
Cinnamaldehyde 98%
Catalog # APOSOR55404-100G
Supplier:  Apollo Scientific
CAS Number:  104-55-2
Cinnamaldehyde 98%

Bottle, 100 g

CAS Number:  104-55-2
Price
21,80 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  trans-Cinnamaldehyde, 3-Phenylacrylaldehyde
  • CAS Number:104-55-2
  • Formula:C₉H₈O
  • MDL Number:MFCD00007000
  • Molecular Weight:132.16 g/mol
  •  :Density: 1.112 g/cm³ (20 °C)
  • Storage Temperature:Freezer
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g

Specifications