0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
4-Hydroxy-2-methoxybenzaldehyde
4-Hydroxy-2-methoxybenzaldehyde 99.5+%
4-Hydroxy-2-methoxybenzaldehyde 99.5+%
4-Hydroxy-2-methoxybenzaldehyde 99.5+%
Bottle, 1 g
APOSOR0857-1G
:
Apollo Scientific
4-Hydroxy-2-methoxybenzaldehyde 99.5+%
APOSOR0857-1G
:
Apollo Scientific
:
18278-34-7
Expand image
4-Hydroxy-2-methoxybenzaldehyde 99.5+%
Bottle, 1 g
Add to cart
:
18278-34-7
21,80 €
Unit of Measure
1 * 1 g
-
+
Add to cart
:
18278-34-7
:
C₈H₈O₃
:
MFCD00051964
:
152.15 g/mol
:
156…161 °C
:
Ambient
Pack type:
Bottle
Pk:
1 g