Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
IGMF 30713 Induktiver Sensor **************************** M18x1x80, 1.4571/PTFE, Sn 5b, 24 V DC, 2-Leiter Öffner, -25 °C...+120 °C, IP68+IP69K, 10 m FEP Kabel [S30713] 1 * 1 ST
Catalog # EGEES30713
Supplier:
EGE ELKTRONIK SPEZIAL SENSOREN
Expand image
IGMF 30713 Induktiver Sensor **************************** M18x1x80, 1.4571/PTFE, Sn 5b, 24 V DC, 2-Leiter Öffner, -25 °C...+120 °C, IP68+IP69K, 10 m FEP Kabel [S30713] 1 * 1 ST
Catalog # EGEES30713
Supplier:
EGE ELKTRONIK SPEZIAL SENSOREN