Order Entry
Export
Orders LinkContactUsLinkComponent
Adapters for universal centrifuges

Adapters for universal centrifuges

Adapters for 50 ml rotor bores or adapters
Catalog # SIMPT330AD50
Supplier:   Simport Scientific
Adapters for universal centrifuges
Catalog # SIMPT330AD50
Supplier:  Simport Scientific
Adapters for universal centrifuges
Adapters for universal centrifuges

Adapters for 50 ml rotor bores or adapters

Price
Inquire For Price
Log in for availability
1 * 8 items
Product Details & Documents

These universal centrifuge adapters are made of acetal plastic.

  • Reusable and freezable (–90 °C)
Specifications
  • Description:
    Adapters for 50 ml rotor bores or adapters