Order Entry
Export
Orders LinkContactUsLinkComponent
Affinity chromatography media, MabSelect™ PrismA

Affinity chromatography media, MabSelect™ PrismA

MabSelect™ PrismA, Liquid, 60 µm, 200 ml
Catalog # 17-5498-02
Supplier:   Cytiva
Affinity chromatography media, MabSelect™ PrismA
Catalog # 17-5498-02
Supplier:  Cytiva
CAS Number:  
Affinity chromatography media, MabSelect™ PrismA
Affinity chromatography media, MabSelect™ PrismA

MabSelect™ PrismA, Liquid, 60 µm, 200 ml

CAS Number:  
Price
5 340,00 €
Log in for availability
1 * 200 mL
Product Details & Documents

MabSelect™ PrismA has been improved with an optimised high-flow agarose base matrix and a genetically engineered protein A-derived ligand, allowing future demands in mAb processing to be met.

  • Enhanced dynamic binding capacity allows high mass throughput of processed mAb per resin volume unit
  • Excellent alkaline stability enables efficient cleaning and sanitisation using 0,5 to 1,0 M NaOH for improved process economy, bioburden, control and robustness
  • Covered by a comprehensive security of supply program, including dual sources of the agarose base matrix and protein A ligand

MabSelect™ PrismA builds on the proven track record of MabSelect™ and MabSelect™ SuRe™ resins in commercial mAb production.

Specifications

Specifications

  • Particle Size:
    60 µm
  • Pk:
    200 ml
  • Description:
    MabSelect™ PrismA
  • Type:
    Liquid
  • Cat. No.:
    17-5498-02
  • Matrix:
    Rigid, highly cross-linked agarose
  • Binding capacity:
    ~ 40 mg polyclonal IgG/ml resin, 2 min residence time<br />~ 80 mg polyclonal IgG/ml resin, 6 min residence time
  • Ligand:
    Alkaline-stabilised protein A-derived (E.coli)
  • pH range:
    3 – 12
  • Ligand coupling method:
    Epoxy
  • Storage temperature:
    2 - 8 °C

Specifications