Empty FPLC columns, G-TRAP™
PP, 5 ml
Catalog # GENO786-1291
Supplier: G-Biosciences
Jump to:
Product Details & Documents
These empty columns are designed for fast protein liquid chromatography.
- Frit material is ultra-high molecular weight polyethylene
Columns are made of polypropylene and come in 1 ml and 5 ml sizes.
Delivery information: Includes column tube, sealing plug, sealing sleeve, top stop plug and bottom stop plug, top (large) frit, bottom (small) frit (inserted in column).
Specifications
- Capacity:5 ml
- Seq:02
- Column hardware:PP
- Cat. No.:GENO786-1291
- Max. pressure:3 bar



