Orders LinkContactUsLinkComponent
VWR® JustCast, Casting chamber, adjustable
VWR® JustCast, Casting chamber, adjustable
JustCast casting tray
Catalog # 700-0849
Supplier:   VWR Collection
VWR® JustCast, Casting chamber, adjustable
Catalog # 700-0849
Supplier:  VWR Collection
VWR® JustCast, Casting chamber, adjustable
VWR® JustCast, Casting chamber, adjustable
JustCast casting tray
Price
540,00 €
Log in for availability
1 * 1 items
Product Details & Documents
The JustCast system is a flexible and quick system for casting multiple PerfectBlue™ compatible 'mini' gels at the same time, without the need to tape. Adjustable to fit all gel trays featured in the PerfectBlue™ mini gel series, this system comes fitted with an easy to read bubble level and adjustable feet to ensure a level casting of the gel. At any one time, the system can fit three Mini S, two Mini M or two Mini L trays.
Specifications
  • Description:
    JustCast casting tray
  • Seq:
    02