ContactUsLinkComponent
NucleoMag DNA Food (4x96) 4 x 96 preps for the isolation of DNA from food and feed NucleoMag B-Beads, buffers, Proteinase K __UN 1 993 Flammable liquid, n.o.s. (ethanol, 2-propanol mixture) 3 III 1.1 L ADR/GGVSEB F1 (E) LQ 1 * 4 items
Catalog # MANA744945.4
Supplier:  MACHEREY-NAGEL
undefined
NucleoMag DNA Food (4x96) 4 x 96 preps for the isolation of DNA from food and feed NucleoMag B-Beads, buffers, Proteinase K __UN 1 993 Flammable liquid, n.o.s. (ethanol, 2-propanol mixture) 3 III 1.1 L ADR/GGVSEB F1 (E) LQ 1 * 4 items
Catalog # MANA744945.4
Supplier:  MACHEREY-NAGEL