ContactUsLinkComponent
NucleoBond Smart Rack light grey rack for use with NucleoBond Xtra Midi, Xtra Maxi, Xtra BAC, AX 20, AX 100, AX 500, AX 2000, and AX 10000 Columns pack of 1 1 * 1 SET
Catalog # MANA740413
Supplier:  MACHEREY-NAGEL
undefined
NucleoBond Smart Rack light grey rack for use with NucleoBond Xtra Midi, Xtra Maxi, Xtra BAC, AX 20, AX 100, AX 500, AX 2000, and AX 10000 Columns pack of 1 1 * 1 SET
Catalog # MANA740413
Supplier:  MACHEREY-NAGEL