ContactUsLinkComponent
RS 232 connection cable 9-pin., female/female, crossed, (2-3/3-2) PC connection of NANOCOLOR 500 D, 400 D, 350 D, 300 D, 250 D, PF-1 1, PF-10, Lumi 10 and NANOCOLOR heating blocks 1 * 1 items
Catalog # MANA919773
Supplier:  MACHEREY-NAGEL
undefined
RS 232 connection cable 9-pin., female/female, crossed, (2-3/3-2) PC connection of NANOCOLOR 500 D, 400 D, 350 D, 300 D, 250 D, PF-1 1, PF-10, Lumi 10 and NANOCOLOR heating blocks 1 * 1 items
Catalog # MANA919773
Supplier:  MACHEREY-NAGEL