Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Grinding bowl stainless steel, with steel casing, 45 ml volume for PULVERISETTE 7 premium line null
Catalog # FRIT50.9720.00
Supplier:
FRITSCH
Expand image
Grinding bowl stainless steel, with steel casing, 45 ml volume for PULVERISETTE 7 premium line null
Catalog # FRIT50.9720.00
Supplier:
FRITSCH