ContactUsLinkComponent
Cartridge with cpl. flow measuring cell incl. insertion block for insertion into the small volume wet dispersion unit SVM - article no. 22.8599.00 for Laser Particle Sizers ANALYSETTE 22 MicroTec plus and NanoTec plus 1 * 1 items
Catalog # FRIT22.8590.04
Supplier:  FRITSCH
Cartridge with cpl. flow measuring cell incl. insertion block for insertion into the small volume wet dispersion unit SVM - article no. 22.8599.00 for Laser Particle Sizers ANALYSETTE 22 MicroTec plus and NanoTec plus 1 * 1 items
Catalog # FRIT22.8590.04
Supplier:  FRITSCH