Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
test sieve 8/2inches mesh = 200 ASTM (0.075 mm) 1 * 1 items
Catalog # FRIT35.5820.03
Supplier:
FRITSCH
Expand image
test sieve 8/2inches mesh = 200 ASTM (0.075 mm) 1 * 1 items
Catalog # FRIT35.5820.03
Supplier:
FRITSCH