Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Set of sieve shells 4 mm round perforation made of stainless steel null
Catalog # FRIT14.4564.00
Supplier:
FRITSCH
Expand image
Set of sieve shells 4 mm round perforation made of stainless steel null
Catalog # FRIT14.4564.00
Supplier:
FRITSCH