Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
test sieve 400/65 mm mesh = 45 = 355 µm - ASTM - E-11-1995 - mesh 1 * 1 items
Catalog # FRIT34.4110.02
Supplier:
FRITSCH
Expand image
test sieve 400/65 mm mesh = 45 = 355 µm - ASTM - E-11-1995 - mesh 1 * 1 items
Catalog # FRIT34.4110.02
Supplier:
FRITSCH