Orders LinkContactUsLinkComponent
Cover glasses
Cover glasses
#1, 5 mm
Catalog # HECH41001105
Cover glasses
Catalog # HECH41001105
Cover glasses
Cover glasses
#1, 5 mm
Price
319,00 €
Log in for availability
1 * 1.000 items
Product Details & Documents
Borosilicate glass highly resistant cover glasses with precise and faultless cut.
Ordering information: Available in different diameters.
Specifications
  • Ø:
    5 mm
  • No.:
    #1
  • Seq:
    02