Order Entry
Export
Orders LinkContactUsLinkComponent
 

Cleaning cloths, MicroClean Plus

Grey, 400×450 mm, 5 cloths/pack
  VILE152534
Cleaning cloths, MicroClean Plus
  VILE152534
Cleaning cloths, MicroClean Plus
Cleaning cloths, MicroClean Plus

Grey, 400×450 mm, 5 cloths/pack

 
621,00 €
   
1 * 100 items
 

MicroClean Plus is a high performance cleaning cloth made from a uniquely fine microfibre (70% polyester and 
30% polyamide). Great streak- and lint-free results are achieved with just one wipe. Ideal for quick dirt removal even without chemicals.

  • Extremly durable
  • Micron Fibre Technology for deeper and more effective cleaning
  • MicroClean dries quickly: Bacteria growth is prevented
  • Washable and reusable: Reduces waste and protects the environment
  • Colour coding system allows a cleaning process according to hygiene standards

MicroClean Plus is suitable for demanding cleaning tasks in general building cleaning and Hotels/Restaurants/Catering on every kind of surface - with or without cleaning agents.

 
  • Packed:
    5 cloths/pack
  • Colour:
    Grey
  • L×W:
    400×450 mm