Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
10ml PrecisionULTRA qPCR Master Mix with ROX at a reduced level (10x 1ml) 1 * 1.000 Tests
Catalog # PULTRA-LR-10ML
Supplier:
PRIMERDESIGN
Expand image
10ml PrecisionULTRA qPCR Master Mix with ROX at a reduced level (10x 1ml) 1 * 1.000 Tests
Catalog # PULTRA-LR-10ML
Supplier:
PRIMERDESIGN