Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
1 ml PrecisionPLUS qPCR Master Mix for capillary LightCycler 1.0-2.0 1 * 100 Tests
Catalog # ZPPLUSCLSY1ML
Supplier:
PRIMERDESIGN
Expand image
1 ml PrecisionPLUS qPCR Master Mix for capillary LightCycler 1.0-2.0 1 * 100 Tests
Catalog # ZPPLUSCLSY1ML
Supplier:
PRIMERDESIGN