Orders LinkContactUsLinkComponent
VWR®, IOS W-Plan Objectives
IOS W-Plan objective
  630-2949
VWR®, IOS W-Plan Objectives
  630-2949
VWR®, IOS W-Plan Objectives
IOS W-Plan objective
46,80 €
1 * 1 items
These high contrast objectives are ideal for routine observations in laboratories.
  • Ensure field flatness up to F.N. 22
  • Based on infinity-corrected optical system
  • 100× is also available with iris for darkfield
  • Magnification:
    4×/0,10
  • Description:
    IOS W-Plan objective
  • Seq:
    151