Orders LinkContactUsLinkComponent
Inspection date labels
Inspection date labels
Inspection date labels, Vinyl, 2028, Green, 15 mm
Catalog # BRDY834118
Supplier:   Brady
Inspection date labels
Catalog # BRDY834118
Supplier:  Brady
Inspection date labels
Inspection date labels
Inspection date labels, Vinyl, 2028, Green, 15 mm
Price
142,00 €
Log in for availability
1 * 600 items
Product Details & Documents
These inspection date labels are made of vinyl cloth. They can be used wherever a frequent and regular inspection of machinery and equipment is required. Simply mark the label with a hole punch and apply wherever you need it.
  • Show evidence of unauthorised removal
Use the yearly colour system to easily identify when your equipment was last inspected.
Specifications
  • Colour:
    Green
  • Description:
    Inspection date labels
  • Dimensions:
    15 mm
  • Material:
    Vinyl
  • Legend:
    2028