Labels, hazard warning, electricity
Hazard labels, Polyester, Black on yellow, 200×173 mm, 1 label/card
Catalog # BRDY250223
Supplier: Brady
Jump to:
Product Details & Documents
These electricity hazard warning labels are made of polyester and are used for prevention of accidents.
- Self-adhesive
- Clear identification of voltage, phases, grounding and insulation
Specifications
- Colour:Black on yellow
- Description:Hazard labels
- Dimensions:200×173 mm
- Material:Polyester
- Seq:03
- Packed:1 label/card