Order Entry
Export
Orders LinkContactUsLinkComponent
Mouse Beta-2-Microglobulin ELISA Kit

Mouse Beta-2-Microglobulin ELISA Kit

Mouse Beta-2-Microglobulin ELISA Kit, 96 tests
Catalog # ABCAAB223590-96
Supplier:   Abcam
Mouse Beta-2-Microglobulin ELISA Kit
Catalog # ABCAAB223590-96
Supplier:  Abcam
CAS Number:  
Mouse Beta-2-Microglobulin ELISA Kit
Mouse Beta-2-Microglobulin ELISA Kit

Mouse Beta-2-Microglobulin ELISA Kit, 96 tests

CAS Number:  
Price
764,00 €
Log in for availability
1 * 96 Tests
Product Details & Documents

Mouse Beta-2-Microglobulin ELISA kit is a single-wash 90-min Simplestep used to quantify Mouse Beta-2-Microglobulin with a sensitivity of 1.165 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader. Different formats for different needs: 384-well plate for higher throughput.

Specifications

Specifications

  • Description:
    Mouse Beta-2-Microglobulin ELISA Kit
  • Tests per kit:
    96 tests
  • Primary antibody reactivity:
    Mouse
  • Sample type:
    Urine, Heparin Plasma, Citrate plasma, Cell culture supernatant, Serum, EDTA Plasma
  • Detection range:
    3,125 - 200 pg/ml
  • Storage temperature:
    4 °C
  • Assay Type:
    Sandwich
  • Time to Results:
    1 hr 30 min
  • Cat. No.:
    ABCAAB223590-96
  • Assay duration:
    One step
  • Detection method:
    Colorimetric
  • Sensitivity:
    1,165 pg/ml
  • Format:
    Pre-coated
  • Target protein:
    beta 2 Microglobulin
  • Regulatory status:
    RUO

Specifications