Order Entry
Export
Orders LinkContactUsLinkComponent
Mouse Recombinant FGF8 (from E. coli)

Mouse Recombinant FGF8 (from E. coli)

Catalog # ABCAAB218083-5
Supplier:   Abcam
Mouse Recombinant FGF8 (from E. coli)
Catalog # ABCAAB218083-5
Supplier:  Abcam
CAS Number:  
Mouse Recombinant FGF8 (from E. coli)
Mouse Recombinant FGF8 (from E. coli)
CAS Number:  
Price
267,00 €
Log in for availability
1 * 5 µG
Product Details & Documents

Recombinant human/mouse FGF8 protein (Active) is a Human Full Length protein, in the 23 to 215 aa range, expressed in Escherichia coli, with >95% purity, <0,05 EU/µg endotoxin level and suitable for SDS-PAGE, FuncS.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications

Specifications

  • Form:
    Lyophilised
  • Protein/peptide name:
    FGF8
  • Protein synonyms:
    AIGF|Fibroblast growth factor 8|FGF-8|Androgen-induced growth factor|Heparin-binding growth factor 8|HBGF-8
  • Purity:
    >95%
  • Source:
    E. coli
  • Biological activity:
    Bioactive
  • Shipping temperature:
    20
  • Cat. No.:
    ABCAAB218083-5
  • Protein/peptide type:
    Recombinant
  • Endotoxin content:
    <0,05 EU/µg
  • Storage conditions:
    Store at –20 °C
  • Formulation:
    Constituents: 0,29% Sodium chloride, 0,08% Sodium phosphate
  • Conjugation:
    Unconjugated
  • Tested applications:
    FuncS, SDS-PAGE
  • Species:
    Mouse
  • Animal-free:
    N

Specifications