ContactUsLinkComponent
Shipping system insulated ChillTherm national 3L 2-8C provides temperature control for up to 24h 1 * 40 items
Catalog # SCATCTN3
Supplier:  Sonoco Thermosafe
Shipping system insulated ChillTherm national 3L 2-8C provides temperature control for up to 24h 1 * 40 items
Catalog # SCATCTN3
Supplier:  Sonoco Thermosafe