Needles replacement parts
Needle seal, SIL-30AC, SIL-30ACMP, 228-52253-00, SHIM
Catalog # CTSA11-4052
Supplier: SCIENCIX
Jump to:
Product Details & Documents
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Sciencix parts are equivalent to the corresponding OEM part.
Specifications
- Description:Needle seal
- Seq:06
- OEM manufacturer:SHIM
- OEM equivalent:228-52253-00
- Model:SIL-30AC, SIL-30ACMP