Orders LinkContactUsLinkComponent
Objectives for high-content cell-imaging system, ImageXpress
Objectives for high-content cell-imaging system, ImageXpress
Fluor objective (spring)
Catalog # MLDV1-6300-0411
Supplier:   Molecular Devices
Objectives for high-content cell-imaging system, ImageXpress
Catalog # MLDV1-6300-0411
Supplier:  Molecular Devices
Objectives for high-content cell-imaging system, ImageXpress
Fluor objective (spring)
Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Magnification:
    20× (0,75 mm)
  • Description:
    Fluor objective (spring)
  • Seq:
    10