Objectives for high-content cell-imaging system, ImageXpress
Fluor objective (spring)
Catalog # MLDV1-6300-0122
Supplier: Molecular Devices
Objectives for high-content cell-imaging system, ImageXpress
Catalog # MLDV1-6300-0122
Supplier: Molecular Devices
Jump to:
Product Details & Documents
Specifications
- Magnification:10× (0,50 mm)
- Description:Fluor objective (spring)
- Seq:9