Orders LinkContactUsLinkComponent
High content cell imaging system, micro confocal, ImageXpress
High content cell imaging system, micro confocal, ImageXpress
Micro confocal transmitted light option with phase contrast includes 20× Ph1 S plan fluor ELWD adm 0,45 N.A objective
Catalog # MLDV5041310
Supplier:   Molecular Devices
High content cell imaging system, micro confocal, ImageXpress
Catalog # MLDV5041310
Supplier:  Molecular Devices
High content cell imaging system, micro confocal, ImageXpress
High content cell imaging system, micro confocal, ImageXpress
Micro confocal transmitted light option with phase contrast includes 20× Ph1 S plan fluor ELWD adm 0,45 N.A objective
Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents
The ImageXpress® Micro-confocal high content imaging system helps to expand the boundaries of research with a combination of speed, sensitivity and flexibility, giving you higher quality images, at a faster throughput, with more powerful analysis.
  • Quickly and easily switch between widefield and several confocal disc configurations, including an exclusive slit option, to capture images at high throughput speeds
  • Image more than 160000 wells per day in confocal mode
  • Excellent image quality via the unique optical path technology
  • Large field of view enables whole-well imaging
  • Tackle a wide range of assays with water, oil and air objectives ranging from 1× to 100×
  • Expand your research capabilities with water immersion objectives, transmitted light, phase contrast optics, on-board liquid handling, and environmental control options
The ImageXpress® Micro-confocal system lets you capture research quality images of a vast range of assays, including whole organisms, thick tissues, 3-D spheroid assays, and cellular or intracellular events - at a speed you would only expect from widefield screening. Benefit from a wide range of imaging modes, including phase contrast and brightfield label-free imaging, fluorescence, widefield, colorimetric, and confocal imaging, to generate publication quality images at high throughput. The system is designed to be exceptionally versatile with user exchangeable filter cubes, a selection of confocal geometries to match your exact assay requirements, on board fluidics and environmental control, the option to benefit from laser illumination, and much more.
Combined with MetaXpress® high content image acquisition and analysis software, the ImageXpress® Micro-confocal system provides you with a complete, customisable, high throughput screening solution to discover your landmark scientific breakthrough.
 : Supplied with a 10× plan-apochromatic 0,45 N.A objective, MetaXpress® imaging and analysis software, Windows® 7 64-bit acquisition computer with license to run MetaXpress® powercore and MDCstore database. Confocal screening option includes eight position filter wheel, five position dichroic wheel and dual disk unit.
Specifications
  • Description:
    Micro confocal transmitted light option with phase contrast includes 20× Ph1 S plan fluor ELWD adm 0,45 N.A objective
  • Seq:
    06
Complementary Products
Product Information
  • Catalog #
    Description
    Pk
    Serum format
    Sterility
    Packed
    Sterile
    Working volume
    Well volume
    Colour
    Recommended working volume
    Availability
    Price
  • 30009-2
    Caspase-3 DEVD-R110 Fluorometric HTS Assay Kit, 10 ml
    1 KIT
    Log in for availability
    1 * 1 KIT - 294,00 €
  • 30026.
    Calcein-AM Cell Viability Assay kit, 1000 assays
    1 KIT
    Log in for availability
    1 * 1 KIT - 417,00 €
  • 100.103.010
    GrowDex®, 10 ml syringe
    1
    Log in for availability
    1 * 1 items - 290,00 €
  • S1810-500
    Foetal bovine serum, South American origin
    Frozen
    15
    Log in for availability
    1 * 500 mL - 330,00 €
  • 30002.
    Viability/Cytotoxicity Assay kit for Animal Live & Dead Cells, 300 assays
    1 KIT
    Log in for availability
    1 * 1 KIT - 440,00 €
  • 30029.
    NucView™ 488 Caspase-3 Assay Kit for live cells, 100 assays
    1 KIT
    Log in for availability
    1 * 1 KIT - 631,00 €
  • 735-0523
    Microlite™ 1+ strip, 1×12 per strip
    Log in for availability
    1 * 320 items - 228,00 €
  • CORN3904BC
    20/bag
    96-well microplate, clear flat bottom, tissue culture treated, with lid
    75 - 200 µl
    21
    360 µl
    Black
    Log in for availability
    1 * 100 items - 1 160,00 €
  • 734-2327
    1/bag
    0,075 - 0,20 ml
    0,33 cm²
    +
    17
    Clear
    Log in for availability
    1 * 100 items - 243,00 €
  • 734-2097
    200 µl
    96-MicroWell™ plate, flat bottomed wells, with lid
    Transparent
    Log in for availability
    1 * 50 items - 200,00 €