High content cell imaging system, ImageXpress Micro 4
Product Details & Documents
The ImageXpress Micro 4 high content imaging system represents the culmination of four generations of imaging expertise. The agile design allows you to boost your research with this faster than ever system, while providing the option to upgrade to confocal in the future to align with your research needs.
- Tackle a wide range of assays with oil and air objectives ranging from 1× to 100×
- Automated five-position filter cube changer to facilitate multiplex assays with up to five filter sets and four objectives for a single experiment
- Optically-encoded linear voice coil driven stages with encoders that measure position with better than 25 nm resolution, to achieve high accuracy and precision in X, Y and Z axes
- Expand your research capabilities with environmental control: Temperature, humidity and CO₂ control for multi-day, live cell experiments), transmitted light true phase contrast, confocal imaging (field upgradable), and fluidics (onboard disposable pipetting)
Capture images of whole organisms and cellular or intracellular events with the ImageXpress Micro 4 system, configured to suit your specific biological needs. We offer unlimited configurability with user exchangeable filter cubes, a wide range of objective lenses and environmental control, transmitted light, confocal imaging and fluidic options. Speed up your research by acquiring more than 245000 wells per day at 4× magnification with the system’s unique multi-well crop feature, and resolve the fine detail you need with increasing magnifications.
Combined with MetaXpress high content image acquisition and analysis software, the ImageXpress Micro 4 system is an end-to-end solution to interpret images, understand data and explore ideas.
A Nuclear Translocation Screening Assay for ß-catenin modulators High-content screening of neuronal toxicity using iPSC-derived human neurons High-throughput confocal imaging of spheroids for screening cancer therapeutics High-throughput imaging assays using zebrafish, a model organism for human disease Intelligent time-lapse imaging to enhance live cell assay development
High Content Screening of Neuronal Toxicity using IPSC derived Human-Neurons High Throughput Imaging Assays using Zebrafish a Model Organism for Human Disease Intelligent Time-lapse Imaging to enhance Live Cell Assay Development Monitoring BNP and Cell Size to determine Hypertrophic in Human Cardiomyocytes Nuclear Translocation Screening Assay for ß-catenin Modulators
Specifications
- Model:ImageXpress Micro 4
- Resolution:25 - 100 nm
- Description:High content imaging system
- Cat. No.:MLDVMICRO4
- Condenser:65 mm WD and PhL, Ph1, and Ph2 selectable phase rings
- W×D×H:530×860×480 mm
- Illumination:Broad spectrum light source − Optional upgrade
- Software:MetaXpress including application module, custom module, and journal
- Magnification:1 – 100×
- Weight:95 kg
- Sensor:High quantum efficiency, 16 bit CMOS
Complementary Products
Product Information
- Catalog #DescriptionPkSerum formatSterilityPackedRecommended working volumeCulture areaSterileColourWorking volumeAvailabilityPrice
- 30026.Calcein-AM Cell Viability Assay kit, 1000 assays1 KITLog in for availability1 * 1 KIT - 417,00 €
- 30029.NucView™ 488 Caspase-3 Assay Kit for live cells, 100 assays1 KITLog in for availability1 * 1 KIT - 631,00 €
- S1810-500Foetal bovine serum, South American originFrozen15Log in for availability1 * 500 mL - 330,00 €
- 30002.Viability/Cytotoxicity Assay kit for Animal Live & Dead Cells, 300 assays1 KITLog in for availability1 * 1 KIT - 440,00 €
- 30009-2Caspase-3 DEVD-R110 Fluorometric HTS Assay Kit, 10 ml1 KITLog in for availability1 * 1 KIT - 294,00 €
- CORN3904BC20/bag96-well microplate, clear flat bottom, tissue culture treated, with lid75 - 200 µl21360 µlBlackLog in for availability1 * 100 items - 1 160,00 €
- 734-2097200 µl96-MicroWell™ plate, flat bottomed wells, with lidTransparentLog in for availability1 * 50 items - 200,00 €



