ContactUsLinkComponent
Set therm. similar Landsberger, -1+102:0,05°C, lower part 180x6-7mm, 6 therm., in stuffed case, null
Catalog # AMARN63970-SET/CC
Supplier:  Amarell
Set therm. similar Landsberger, -1+102:0,05°C, lower part 180x6-7mm, 6 therm., in stuffed case, null
Catalog # AMARN63970-SET/CC
Supplier:  Amarell