Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Amarell-Special-Therm., +248+302:0,1°C, enclosed scale, cap. round, colourless, gallium filled, c.450x9,0-9,5mm, Richter-top finish, with ice point scale, suitable for government calibration 1 * 1 items
AMARN61630-GA
:
Amarell
Amarell-Special-Therm., +248+302:0,1°C, enclosed scale, cap. round, colourless, gallium filled, c.450x9,0-9,5mm, Richter-top finish, with ice point scale, suitable for government calibration 1 * 1 items
AMARN61630-GA
:
Amarell