ContactUsLinkComponent
Anschütztherm., +248+302:0,1°C, solid stem, white backed, gallium filled, 340x5-6mm, total immersion, with button at top, suitable f or government calibration 1 * 1 items
Catalog # AMARN60082-GA
Supplier:  Amarell
Anschütztherm., +248+302:0,1°C, solid stem, white backed, gallium filled, 340x5-6mm, total immersion, with button at top, suitable f or government calibration 1 * 1 items
Catalog # AMARN60082-GA
Supplier:  Amarell