ContactUsLinkComponent
Anschütztherm., -10+52:0,1°C, solid stem, white backed, capillary tube specially coated inside, with non-wetting blue special liqui d, 340x5-6mm, total immersion, with button at top, null
Catalog # AMARN60072-CB
Supplier:  Amarell
Anschütztherm., -10+52:0,1°C, solid stem, white backed, capillary tube specially coated inside, with non-wetting blue special liqui d, 340x5-6mm, total immersion, with button at top, null
Catalog # AMARN60072-CB
Supplier:  Amarell