ContactUsLinkComponent
Anschütztherm., +298+355:0,2°C, enclosed scale, cap. lens front, colourless, gallium filled, 175x5-6mm, total immersion, Richter-top finish, suitable for government calibration 1 * 1 items
Catalog # AMARN60034-GA
Supplier:  Amarell
Anschütztherm., +298+355:0,2°C, enclosed scale, cap. lens front, colourless, gallium filled, 175x5-6mm, total immersion, Richter-top finish, suitable for government calibration 1 * 1 items
Catalog # AMARN60034-GA
Supplier:  Amarell