ContactUsLinkComponent
Crystallization point therm., similar DIN 12785, +4,0+6,0:0,01°C, enclosed scale, capillary tube specially coated inside, with non- wetting blue special liquid, total immersion, 250x6mm, Richter-top finish without button, null
Catalog # AMARM253600-CB
Supplier:  Amarell
Crystallization point therm., similar DIN 12785, +4,0+6,0:0,01°C, enclosed scale, capillary tube specially coated inside, with non- wetting blue special liquid, total immersion, 250x6mm, Richter-top finish without button, null
Catalog # AMARM253600-CB
Supplier:  Amarell