ContactUsLinkComponent
Melting point therm., similar Lindström, 0+360:1°C, enclosed scale, cap. lens front, blue reflecting, gallium filled, 340x5-6mm, imm ersion 50mm, Richter-top finish with button, suitable for government verification 1 * 1 items
Catalog # AMARM250460-GA
Supplier:  Amarell
Melting point therm., similar Lindström, 0+360:1°C, enclosed scale, cap. lens front, blue reflecting, gallium filled, 340x5-6mm, imm ersion 50mm, Richter-top finish with button, suitable for government verification 1 * 1 items
Catalog # AMARM250460-GA
Supplier:  Amarell