ContactUsLinkComponent
Melting point therm., +200+360:0,5°C, enclosed scale, cap. prismatic, colourless, gallium filled, 250x6-7mm, immersion 30mm, Richter -top finish with button, suitable for government verification 1 * 1 items
Catalog # AMARM250404-GA
Supplier:  Amarell
Melting point therm., +200+360:0,5°C, enclosed scale, cap. prismatic, colourless, gallium filled, 250x6-7mm, immersion 30mm, Richter -top finish with button, suitable for government verification 1 * 1 items
Catalog # AMARM250404-GA
Supplier:  Amarell