ContactUsLinkComponent
Therm. straight stem, similar DIN 16174, 0+100:1°C, cap. prismatic, yellow backed, milk glass scale, red special liquid, upper part 235x20,5mm, lower part 303x8mm, immersion 228mm, cork-glass-top finish 1 * 1 items
Catalog # AMART91150-FL
Supplier:  Amarell
Therm. straight stem, similar DIN 16174, 0+100:1°C, cap. prismatic, yellow backed, milk glass scale, red special liquid, upper part 235x20,5mm, lower part 303x8mm, immersion 228mm, cork-glass-top finish 1 * 1 items
Catalog # AMART91150-FL
Supplier:  Amarell