ContactUsLinkComponent
Therm. straight stem, DIN 16174, -60+40:1C, cap. prismatic, yellow backed, milk glass scale, toluene filled, upper part 235x20,5mm, lower part 165x8mm, immersion 138mm, cork-glass-top finish 1 * 1 items
Catalog # AMART91060-TOL
Supplier:  Amarell
Therm. straight stem, DIN 16174, -60+40:1C, cap. prismatic, yellow backed, milk glass scale, toluene filled, upper part 235x20,5mm, lower part 165x8mm, immersion 138mm, cork-glass-top finish 1 * 1 items
Catalog # AMART91060-TOL
Supplier:  Amarell