ContactUsLinkComponent
Therm. straight stem, similar DIN 16174, 0+600:5°C, cap. prismatic, colourless, milk glass scale, gallium filled, upper part 235x20, 5mm, lower part 165x8mm, immersion 138mm, cork-glass-top finish 1 * 1 items
Catalog # AMART91460-GA
Supplier:  Amarell
Therm. straight stem, similar DIN 16174, 0+600:5°C, cap. prismatic, colourless, milk glass scale, gallium filled, upper part 235x20, 5mm, lower part 165x8mm, immersion 138mm, cork-glass-top finish 1 * 1 items
Catalog # AMART91460-GA
Supplier:  Amarell