ContactUsLinkComponent
Therm. straight stem, similar DIN 16174, 0+160:2°C, cap. prismatic, yellow backed, milk glass scale, red special liquid, upper part 235x20,5mm, lower part 213x8mm, immersion 138mm, cork-glass-top finish 1 * 1 items
Catalog # AMART91194-FL
Supplier:  Amarell
Therm. straight stem, similar DIN 16174, 0+160:2°C, cap. prismatic, yellow backed, milk glass scale, red special liquid, upper part 235x20,5mm, lower part 213x8mm, immersion 138mm, cork-glass-top finish 1 * 1 items
Catalog # AMART91194-FL
Supplier:  Amarell