ContactUsLinkComponent
Gallium-Quartz-glass therm., 0+1000:5°C, 450x8-9mm, cap. colourless, strip bearing the scale: quartz, total immersion, pin-top finis h with button, suitable for government verification 1 * 1 items
Catalog # AMARQ140050-GA
Supplier:  Amarell
Gallium-Quartz-glass therm., 0+1000:5°C, 450x8-9mm, cap. colourless, strip bearing the scale: quartz, total immersion, pin-top finis h with button, suitable for government verification 1 * 1 items
Catalog # AMARQ140050-GA
Supplier:  Amarell